Caffeic acid phenylethyl ester ≥97% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: (E)-Phenethyl 3-(3,4-dihydroxyphenyl)acrylate

270-244-M010 270-244-M050
89147-474EA 47.96 USD
89147-474 89147-476
Caffeic acid phenylethyl ester ≥97% (by HPLC)
Caffeic acid phenylethyl ester

HIV-1 integrase inhibitor


Active component of propolis from honeybee hives, known to have anti-mitogenic, anti-carcinogenic, anti-inflammatory, and immunomodulatory properties. Potent and specific inhibitor of NF-kappaB activation, induced by TNF-alpha (Prod. No. ALX-522-008 (human) or Prod. No. ALX-522-009 (mouse)), PMA (Prod. No. BML-PE160) and other inflammatory agents. Exhibits inhibitory activity against HIV-1 integrase (IC50=7µM). Suppresses lipid peroxidation and inhibits ornithine decarboxylase, protein tyrosine kinase and lipoxygenase activities. Induces apoptosis.


Synthetic.

Formula: C₁₇H₁₆O₄
MDL Number: MFCD00866470
CAS Number: 115610-29-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=537f37d275d606e2ed8a0134079ff3efbc07976a&vid=6894938b653cb067a76c6d45a42ee5f3fd1bfdbb&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR