Anti-HSF1 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
ADI-SPA-902-D ADI-SPA-902-F
89142-098EA 294 USD
89142-098 89142-100
Anti-HSF1 Rabbit Polyclonal Antibody
Antibodies
HSFs (Heat Shock family of transcription factors), which consists of HSF 1-4, bind to highly conserved Heat shock elements (HSEs) in the promoter regions of heat shock genes, ultimately regulating the expression of Heat shock proteins (Hsps). On exposure to heat shock and other stresses, HSF1 localizes within seconds to discrete nuclear granules and on recovery from stress, HSF1 rapidly dissipates from the stress granules to a diffuse nucleoplasmic distribution.

  • For Western Blot

Recognizes human HSF1. Predicted MW of ~60kDa. Detects a band of 85-95kDa by Western blot depending on hyperphosphorylation of the native protein.

Type: Primary
Antigen: HSF1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR