Anti-DNAK Mouse Monoclonal Antibody [clone: 8E2/2]

Supplier: Enzo Life Sciences
ADI-SPA-880-F ADI-SPA-880-D ADI-SPA-880-J
200062-784EA 607.13 USD
200062-784 200062-782 95043-760
Anti-DNAK Mouse Monoclonal Antibody [clone: 8E2/2]
Antibodies
DnaK possesses an autophosphorylation activity and a weak 5-nucleotidase activity, cleaving the 5 phosphate groups from both ribose and deoxyribose nucleotides. The role of DnaK in ATP-dependent protein-protein interactions has been extended to normal E. coli physiology, where, like eukaryotic Hsp70 homologs, it is thought to participate in the assembly/disassembly of protein complexes.

  • For Western Blot

Recognizes bacteria DnaK. Detects a band of ~70kDa by Western blot.

Type: Primary
Antigen: DNAK
Clonality: Monoclonal
Clone: 8E2/2
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR