Anti-ERP57 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
ADI-SPA-585-D ADI-SPA-585-F
89142-054EA 238.71 USD
89142-054 89142-056
Anti-ERP57 Rabbit Polyclonal Antibody
Antibodies
ERp57 (also known as Grp58), a member of the Thioredoxin superfamily, is an ER protein with disulfide isomerase activity. It has been shown to increase after oncogenic transformation and to be a component of the MHC Class I peptide-loading complex.

  • For Western Blot

Recognizes human, mouse, rat, bovine, dog, guinea pig, hamster, monkey, pig and rabbit ERp57. Detects a band of ~57kDa by Western blot.

Type: Primary
Antigen: ERP57
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR