Anti-MT Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
ADI-SPA-550-D ADI-SPA-550-F
95043-874EA 286.67 USD
95043-874 95043-876
Anti-MT Mouse Monoclonal Antibody
Antibodies
Metallothioneins (MTs) are a class of ubiquitous, low molecular weight, cysteine-rich, metal binding proteins that function in the metabolism of zinc and copper. MT protects against exposure to toxic metals such as mercury and cadmium as well as a variety of other stress conditions. There are four mammalian MT isoforms: MT-I, -II, -III, -IV, each of which is encoded by individual genes. MT expression is induced by heavy metal cations, starvation, elevated temperature, lipopolysaccharide, inflammatory cytokines, free radicals, and ionizing radiation.

  • For ELISA, Flow Cytometry, Western Blot

Recognizes human, mouse, rat, fish, horse and rabbit metallothionein. Detects a band of ~15kDa by Western blot.

Type: Primary
Antigen: MT
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR