IL-10 (Human), ELISA kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-900-036 ADI-901-036
89141-092EA 691.06 USD
89141-092 89141-298
IL-10 (Human), ELISA kit, Enzo Life Sciences
Assays ELISAs
The IL-10 (human), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in < 3 hours
For the quantitative determination of human IL-10 in culture supernatants, plasma, and serum.
Absorbance is read at 450 nm. Ready-to-use liquid color-coded reagents reduce laboratory errors.
Información de suministro: Kit includes microtiter plate, conjugate, antibody, assay buffer 13 concentrate, wash buffer concentrate, standard, TMB substrate, stop solution 2