5(6)-TAMRA

Supplier: BACHEM AMERICAS INC

Synonyms: TAMRA 5 and 6-isomer, Tetramethylrhodamine-5-(and-6)-maleimide, 5(6)-Carboxy-tetramethylrhodamine, 9-[2,4(or 2,5)-Dicarboxyphenyl]-3,6-bis(dimethylamino)-xanthylium inner salt

4045146.0100 4045146.0500
Q-2680.0100BAEA 389.89 USD
Q-2680.0100BA Q-2680.0500BA
5(6)-TAMRA
5(6)-TAMRA

Sequence: 5(6)-Carboxy-tetramethylrhodamine
Synonym(s): 5(6)-TAMRA


Bachem also offers the coumarin derivative Fmoc-3-(7-methoxy-coumarin-4-yl)-Ala-OH (B-3740), the (7-Hydroxycoumarin-4-yl)ethylglycine derivatives B-4205 and F-4270, and a range of Nε-labeled lysine derivatives (please see the subfamily "Nε-Labeled Lysine").


Mixture of 5- and 6-TAMRA (TMR). Often used fluorophore for labeling peptides and proteins. CAS Number (spiro form) 50347-56-1.

Formula: C₂₅H₂₂N₂O₅
Storage Temperature: Freezer
MDL Number: MFCD16661194
CAS Number: 98181-63-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR