EEDQ (ethyl-N-(2-ethoxy-1,2-dihydroquinoline)carboxylate)

Supplier: BACHEM AMERICAS INC

Synonyms: 2-Ethoxy-1-ethoxycarbonyl-1,2-dihydroquinoline, N-Ethoxycarbonyl-2-ethoxy-1,2-dihydroquinoline, Ethyl 2-ethoxyquinoline-1(2H)-carboxylate

4015690.0100 4015690.0500
Q-1735.0100BAEA 1013.8 USD
Q-1735.0100BA Q-1735.0500BA
EEDQ (ethyl-N-(2-ethoxy-1,2-dihydroquinoline)carboxylate)
EEDQ (ethyl-N-(2-ethoxy-1,2-dihydroquinoline)carboxylate)

Sequence: N-Ethoxycarbonyl-2-ethoxy-1,2-dihydroquinoline


EEDQ permits the coupling of acylamino acids with amino acid esters in high yield without concomitant racemization. In a study aiming at the optimization of the coupling of 3,5-dinitrobenzoyl-Leu to 3-aminopropyl-silica (Yang et al.) significant levels of racemization were observed with all the tested coupling reagents except for EEDQ .

Formula: C₁₄H₁₇NO₃
MW: 247.29 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00006703
CAS Number: 16357-59-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR