(S)-2-Hydroxy-3-phenylpropanoic acid

Supplier: BACHEM AMERICAS INC

Synonyms: L-β-Phenyllactic acid, (S)-(-)-3-Phenyllactic Acid, L-3-Phenyllactic Acid, L(-)-3-Phenyllactic acid, (S)-(-)-2-Hydroxy-3-phenylpropionic Acid, L-(-)-3-Phenyllactic acid

4001139.0025 4001139.0100
Q-1375.0025BAEA 196.08 USD
Q-1375.0025BA Q-1375.0100BA
(S)-2-Hydroxy-3-phenylpropanoic acid
(S)-2-Hydroxy-3-phenylpropanoic acid

Sequence: L-β-Phenyllactic acid


Useful chiral synthons, building blocks for depsipeptides.


Synonym(s): (S)-2-Hydroxy-3-phenyl-propionic acid

Formula: C₉H₁₀O₃
Storage Temperature: Refrigerator
MDL Number: MFCD00004244
CAS Number: 20312-36-1
EINECS: 243-726-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR