4-(2-Aminoethyl)-2-methoxyphenol hydrochloride

Supplier: BACHEM AMERICAS INC

Synonyms: 3-Methoxytyramine·HCl, 3-O-Methyldopamine hydrochloride

4011849.0001 4011849.0005
Q-1330.0001BAEA 167.37 USD
Q-1330.0001BA Q-1330.0005BA
4-(2-Aminoethyl)-2-methoxyphenol hydrochloride
4-(2-Aminoethyl)-2-methoxyphenol hydrochloride

Sequence: 3-Methoxytyramine · HCl
Synonym(s): 3-MT · HCl#3-O-Methyldopamine · HCl


A major metabolite of L-DOPA. Elevated concentrations of 3-MT in body fluids are indicators for a variety of brain and carcinoid tumors. N-Acetylation of 3-MT yields NAMDA, which in vitro inhibits GTP cyclohydrolase I, the rate-limiting enzyme for tetrahydrobiopterin biosynthesis. NAMDA also reduces nitrite accumulation, an oxidative metabolite of NO, without directly inhibiting NOS activity. CAS Number (net): 554-52-9.

Formula: C₉H₁₄ClNO₂
MW: 203.666 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00012902
CAS Number: 1477-68-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR