7-Amino-4-methylcoumarin

Supplier: BACHEM AMERICAS INC

Synonyms: 7-Amino-4-methyl-2-benzopyrone, Coumarin 440, Coumarin 120, AMC

4001606.0001 4001606.0005
Q-1025.0001BAEA 206.12 USD
Q-1025.0001BA Q-1025.0005BA
7-Amino-4-methylcoumarin
7-Amino-4-methylcoumarin

Sequence: 7-Amino-4-methylcoumarin
Synonym(s): AMC


Bachem also offers the coumarin derivative Fmoc-3-(7-methoxy-coumarin-4-yl)-Ala-OH (B-3740), the (7-Hydroxycoumarin-4-yl)ethylglycine derivatives B-4205 and F-4270, and a range of Nε-labeled lysine derivatives (please see the subfamily "Nε-Labeled Lysine").


Fluorophore (λex 365 nm, λem 440 nm, in ethanol) for the synthesis of fluorogenic peptidase substrates. Reference compound for assays employing AMC substrates.

Formula: C₁₀H₉NO₂
MW: 175.19 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00006868
CAS Number: 26093-31-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR