Edelfosine

Supplier: BACHEM AMERICAS INC

Synonyms: 1-O-Octadecyl-2-O-methyl-rac-glycero-3-phosphocholine

4030361.0025 4030361.0100
O-1415.0025BAEA 510.11 USD
O-1415.0025BA O-1415.0100BA
Edelfosine
Edelfosine

Sequence: 1-O-Octadecyl-2-O-methyl-rac-glycero-3-phosphocholine
Synonym(s): Edelfosine#ALP#ET 18-OCH3#ET-18-OMe#NSC 324368#NSC 343649#Ro 14-5243


1-O-Octadecyl-2-O-methyl-rac-glycero-3-phosphorylcholine (O-1415) and octadecylphosphorylcholine are efficient anticancer agents. However, as the choline derivatives are rather toxic, homologs have been synthesized in which the nitrogen atom of choline has been exchanged for a phosphorus atom. The resulting compounds were less toxic and retained the cytotoxic activity.


Edelfosine showed an antimetastatic effect in experimental tumors. Moreover, according to the results of Varela et al., the phospholipid could find use as an orally administered antileishmanial drug. Our product O-1385 corresponds to a pure enantiomer of edelfosine.

Formula: C₂₇H₅₈NO₆P
MW: 523.736 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00135190
CAS Number: 70641-51-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR