1-O-Hexadecyl-sn-glycero-3-phosphocholine

Supplier: BACHEM AMERICAS INC

4008363.0025 4008363.0100
O-1300.0025BAEA 245.07 USD
O-1300.0025BA O-1300.0100BA
1-O-Hexadecyl-sn-glycero-3-phosphocholine
1-O-Hexadecyl-sn-glycero-3-phosphocholine

Sequence: 1-O-Hexadecyl-sn-glycero-3-phosphocholine


O-1270 corresponds to PAF (C₁₆), O-1355 to PAF (C₁₈).


Synonym(s): Lyso-PAF (C₁₆)

Formula: C₂₄H₅₂NO₆P
Storage Temperature: Freezer
MDL Number: MFCD00042932
CAS Number: 52691-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR