Z-Val-Arg-Pro-DL-Arg-FluoromethylKetone Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4055669.0500
4055669.1000
N-2010.0500BAEA
441.71
USD
N-2010.0500BA
N-2010.1000BA
Z-Val-Arg-Pro-DL-Arg-FluoromethylKetone Trifluoroacetate
Proteins and Peptides
Cell-permeable, irreversible inhibitor of paracaspase MALT1. The inhibitor was based on the sequence of a published optimal tetrapeptide substrate for the A. thaliana metacaspase AtMC9.