B581

Supplier: BACHEM AMERICAS INC

Synonyms: H-Cys-psi(CH₂NH)Val-psi(CH₂NH)Phe-Met-OH

4025336.0005 4025336.0025
N-1390.0005BAEA 718.94 USD
N-1390.0005BA N-1390.0025BA
B581
B581

Sequence: H-Cys-psi(CH₂NH)Val-psi(CH₂NH)Phe-Met-OH
Synonym(s): B581#Farnesyltransferase inhibitor#N-[(S)-2-((R)-2-Amino-3-mercapto-propylamino)-3-methyl-butyl]-Phe-Met-OH


B581, Cys-psi(CH₂NH)-Val- psi[CH₂NH]-Phe-Met is a farnesyltransferase (FTase) inhibitor of special stability and membrane permeability. Due to its high selectivity, B581 is more than 30-fold more active against FTase (IC₅₀ = 21 nM in vitro) than against geranylgeranyltransferase (GGTase). Inhibitors of the farnesyltransferase are potential candidates for the development of anticancer drugs.

Formula: C₂₂H₃₈N₄O₃S₂
Storage Temperature: Freezer
MDL Number: MFCD18910841
CAS Number: 149759-96-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR