DL-Thiorphan

Supplier: BACHEM AMERICAS INC

4010438.0025 4010438.0100
N-1195.0025BAEA 408.5 USD
N-1195.0025BA N-1195.0100BA
DL-Thiorphan
DL-Thiorphan

Sequence: ((RS)-2-Benzyl-3-mercaptopropionyl)-Gly-OH


Bachem additionally offers deuterated glycine: Fmoc-[D₂]Gly-OH B-4125. For N-methylglycine derivatives see Sarcosine.
See also Boc-Gly-ONp (A-1740) and Z-Gly-ONp (M-1250).


Synonym(s): (DL-3-Mercapto-2-benzylpropanoyl)-Gly-OH

Formula: C₁₂H₁₅NO₃S
Storage Temperature: Freezer
MDL Number: MFCD00058435
CAS Number: 76721-89-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR