Glycyl-L-glutamine
Supplier: BACHEM AMERICAS INC
4001242.0005
4001242.0025
N-1070.0005BAEA
326.8
USD
N-1070.0005BA
N-1070.0025BA
Glycyl-L-glutamine
Glycyl-L-glutamine
Sequence: H-Gly-Gln-OH
For other opioid peptides see, the product families ‘Deltorphins and Dermorphins’, ‘Dynorphin, Analogs and Sequences, ‘Endorphins, MPF, Neoendorphins’, and ‘Opioid Peptides’.
Synonym(s): β-Lipotropin (90-91)
Formula:
C₇H₁₃N₃O₄ Storage Temperature: Freezer |
MDL Number:
MFCD00077124 CAS Number: 13115-71-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...