Ac-Ile-Tyr-Gly-Glu-Phe-NH₂
Supplier: BACHEM AMERICAS INC
4026672.0005 4026672.0025
M-2165.0005BAEA 163.38 USD
M-2165.0005BA M-2165.0025BA
Ac-Ile-Tyr-Gly-Glu-Phe-NH₂
Proteins and Peptides
Ac-IYGEF-amide, excellent small peptide substrate for the protein tyrosine kinase pp60c-src (Km= 368 µM and Vmax= 1.02 µmol min⁻¹ mg⁻¹).