FA-Phe-Phe-OH
Supplier: BACHEM AMERICAS INC
4011445.0100 4011445.0250
M-1760.0100BAEA 317.99 USD
M-1760.0100BA M-1760.0250BA
FA-Phe-Phe-OH
Proteins and Peptides
The FA-peptides M-1400 and M-1410 are common ACE substrates and thus filed in the corresponding product family. For our 3-(2-furyl)acryloyl-amino acids (FA-amino acids) and the labeling reagent FA-OSu (Q-1245), please see the Amino Acids section.
This substrate for a sensitive spectrophotometric assay of serum carboxypeptidase A has also been used in kinetic studies with carboxypeptidase Y.