(Trp⁴)-Kemptide
Supplier: BACHEM AMERICAS INC
4006462.0005 4006462.0025
M-1525.0005BAEA 332.32 USD
M-1525.0005BA M-1525.0025BA
(Trp⁴)-Kemptide
Proteins and Peptides
Chocktide ((Trp⁴)-Kemptide) is readily phosphorylated by cAMP-dependent protein kinase. The phosphorylated peptide, in turn, is an effective substrate for phosphoprotein phosphatase. The changes in the substrate fluorescence intensity as a function of its phosphorylation state provide a sensitive assay of both enzyme activities.