FA-Phe-Gly-Gly-OH
Supplier: BACHEM AMERICAS INC
4003634.0250 4003634.1000
M-1400.0250BAEA 408.5 USD
M-1400.0250BA M-1400.1000BA
FA-Phe-Gly-Gly-OH
Proteins and Peptides
Substrate for a continuous spectrophotometric assay of human angiotensin I-converting enzyme (ACE). Enzymatic cleavage yields FA-Phe-OH (M-1390) and H-Gly-Gly-OH (G-2090).