FA-Gly-Leu-NH₂
Supplier: BACHEM AMERICAS INC
4003615.0250 4003615.1000
M-1375.0250BAEA 334.97 USD
M-1375.0250BA M-1375.1000BA
FA-Gly-Leu-NH₂
Proteins and Peptides
The FA-peptides M-1400 and M-1410 are common ACE substrates and thus filed in the corresponding product family. For our 3-(2-furyl)acryloyl-amino acids (FA-amino acids) and the labeling reagent FA-OSu (Q-1245), please see the Amino Acids section.
A good substrate for thermolysin, the Bacillus subtilis neutral protease, and the fibrinolytic neutral metalloendopeptidase EM 19000 from Streptococcus faecalis.