Z-Gly-Pro-Gly-Gly-Pro-Ala-OH
Supplier: BACHEM AMERICAS INC
Synonyms:
Z-Gly-Pro-Gly-Gly-Pro-Ala-OH
4002149.0100
4002149.0500
M-1260.0100BAEA
117.28
USD
M-1260.0100BA
M-1260.0500BA
Z-Gly-Pro-Gly-Gly-Pro-Ala-OH
Z-Gly-Pro-Gly-Gly-Pro-Ala-OH
Sequence: Z-Gly-Pro-Gly-Gly-Pro-Ala-OH
Substrate for collagenase. Enzymatic cleavage of Z-GPGGPA yields Gly-Pro-Ala (H-3615), which is determined colorimetrically by reaction with ninhydrin.
Formula:
C₂₇H₃₆N₆O₉ Storage Temperature: Freezer |
MDL Number:
MFCD00077021 CAS Number: 13075-38-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...