Suc-Phe-pNA
Supplier: BACHEM AMERICAS INC
4000089.0001
4000089.0005
L-1420.0001BAEA
140.17
USD
L-1420.0001BA
L-1420.0005BA
Suc-Phe-pNA
Proteins and Peptides
Bachem additionally offers deuterated phenylalanine: Fmoc-[ring-D₅]Phe-OH B-4115.
See also Z-Phe-OH (C-2310) and Z-Phe-ONp (C-2345).
Substrate for chymotrypsin, also used for the identification of this enzyme by enzymoblotting.