L-Lysine 4-nitroanilide dihydrobromide

Supplier: BACHEM AMERICAS INC

Synonyms: H-Lys-pNA·2 HBr

4003415.0001 4003415.0005
L-1315.0001BAEA 195.99 USD
L-1315.0001BA L-1315.0005BA
L-Lysine 4-nitroanilide dihydrobromide
L-Lysine 4-nitroanilide dihydrobromide

Sequence: H-Lys-pNA · 2 HBr


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".
See also Z-Lys-ONp · HCl, M-1295.

Formula: C₁₂H₂₀Br₂N₄O₃
Storage Temperature: Freezer
MDL Number: MFCD00050686
CAS Number: 40492-96-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR