H-Ala-Ala-Ala-Pna·HCl

Supplier: BACHEM AMERICAS INC

4000901.0250 4000901.1000
L-1090.0250BAEA 361.17 USD
L-1090.0250BA L-1090.1000BA
H-Ala-Ala-Ala-Pna·HCl
H-Ala-Ala-Ala-Pna·HCl

Sequence: H-Ala-Ala-Ala-pNA · HCl


AAA-pNA, chromogenic substrate for porcine pancreatic elastase and for astacin, a crayfish zinc-endopeptidase.

Formula: C₁₅H₂₂ClN₅O₅
Storage Temperature: Freezer
MDL Number: MFCD00058291
CAS Number: 50450-80-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR