L-Argininamide,L-arginyl-N-2-naphthalenyl, trihydrochloride (9CI)
Supplier: BACHEM AMERICAS INC
Synonyms:
H-Arg-Arg-βNA·3 HCl
4000098.0050
4000098.0250
K-1085.0050BAEA
114.5
USD
K-1085.0050BA
K-1085.0250BA
L-Argininamide,L-arginyl-N-2-naphthalenyl, trihydrochloride (9CI)
L-Argininamide,L-arginyl-N-2-naphthalenyl, trihydrochloride (9CI)
Sequence: H-Arg-Arg-βNA · 3 HCl
RR-βNA, specific substrate for dipeptidyl peptidase III.
Formula:
C₂₂H₃₆Cl₃N₉O₂ MW: 564.941 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00058045 CAS Number: 100900-26-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...