Z-Val-Ala-Asn-AMC
Supplier: BACHEM AMERICAS INC
4029129.0050
4029129.0250
I-1840.0050BAEA
361.17
USD
I-1840.0050BA
I-1840.0250BA
Z-Val-Ala-Asn-AMC
Proteins and Peptides
Substrate for the determination of a new cysteine endopeptidase which is required for antigen processing. Z-VAN-AMC has been used as a fluorogenic substrate for porcine asparaginyl endopeptidase (AEP, legumain).