H-Asn-AMC
Supplier: BACHEM AMERICAS INC
4012553.0250
4012553.1000
I-1435.0250BAEA
1306.82
USD
I-1435.0250BA
I-1435.1000BA
H-Asn-AMC
H-Asn-AMC
Sequence: H-Asn-AMC
For isoasparagine derivatives please see the subfamily "Isoasparagine" filed under "Aspartic Acid".
See also A-1230 and E-3705.
Formula:
C₁₄H₁₅N₃O₄ Storage Temperature: Freezer |
MDL Number:
MFCD00078916 CAS Number: 115047-89-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...