H-Ala-Ala-Phe-AMC (free base)
Supplier: BACHEM AMERICAS INC
4002571.0050
4002571.0250
I-1415.0050BAEA
167.52
USD
I-1415.0050BA
I-1415.0250BA
H-Ala-Ala-Phe-AMC (free base)
Proteins and Peptides
AAF-AMC, fluorogenic substrate for tripeptidyl peptidases I and II and for tripeptide aminopeptidase EC 3.4.11.4.