Suc-Ala-Pro-Ala-AMC
Supplier: BACHEM AMERICAS INC
4005965.0050
4005965.0250
I-1335.0050BAEA
883.44
USD
I-1335.0050BA
I-1335.0250BA
Suc-Ala-Pro-Ala-AMC
Suc-Ala-Pro-Ala-AMC
Sequence: Suc-Ala-Pro-Ala-AMC
See also C-1040, C-2345, I-1000, and L-1435.
Suc-APA-AMC, a sensitive substrate for human leukocyte elastase.
Formula:
C₂₅H₃₀N₄O₈ Storage Temperature: Freezer |
MDL Number:
MFCD00038655 CAS Number: 88467-44-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...