H-Phe-AMC

Supplier: BACHEM AMERICAS INC

4001559.0050 4001559.0250 4001559.1000
I-1285.0050BAEA 114.5 USD
I-1285.0050BA I-1285.0250BA I-1285.1000BA
H-Phe-AMC
H-Phe-AMC

Sequence: H-Phe-AMC


Bachem additionally offers deuterated phenylalanine: Fmoc-[ring-D₅]Phe-OH B-4115.
See also Z-Phe-OH (C-2310) and Z-Phe-ONp (C-2345).

Formula: C₁₉H₁₈N₂O₃
Storage Temperature: Freezer
MDL Number: MFCD00167024
CAS Number: 98516-72-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR