H-Lys-Ala-AMC
Supplier: BACHEM AMERICAS INC
4007641.0050
4007641.0250
I-1260.0050BAEA
167.52
USD
I-1260.0050BA
I-1260.0250BA
H-Lys-Ala-AMC
Proteins and Peptides
KA-AMC, highly sensitive, fluorogenic substrate for dipeptidyl aminopeptidase II (DPP II) and Potent substrate for P. endodontalis dipeptidylpeptidase V (DPP 5).