(S)-2-Amino-4-(7-hydroxy-2-oxo-2H-chromen-4-yl)butanoic acid

Supplier: BACHEM AMERICAS INC

Synonyms: H-4-(7-Hydroxy-4-coumarinyl)-Abu-OH, 7-HCAA

4059541.0100 4059541.0250 4059541.1000
F-4270.0100BAEA 158.5 USD
F-4270.0100BA F-4270.0250BA F-4270.1000BA
(S)-2-Amino-4-(7-hydroxy-2-oxo-2H-chromen-4-yl)butanoic acid
(S)-2-Amino-4-(7-hydroxy-2-oxo-2H-chromen-4-yl)butanoic acid

Sequence: H-4-(7-Hydroxy-4-coumarinyl)-Abu-OH
Synonym(s): H-(7-Hydroxycoumarin-4-yl)-ethyl-Gly-OH#H-(Umbellifer-4-yl)-ethyl-Gly-OH


L-(7-hydroxycoumarin-4-yl)ethylglycine (umbelliferylethylglycine) has been incorporated into proteins as a fluorescent tag.

Formula: C₁₃H₁₃NO₅
MW: 263.249 g/mol
Storage Temperature: Freezer
MDL Number: MFCD22377747
CAS Number: 905442-42-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR