ɑ-Methyl-L-proline
Supplier: BACHEM AMERICAS INC
Synonyms:
(S)-2-Methylpyrrolidine-2-carboxylic acid, (S)-2-Methylproline, H-α-Me-Pro-OH, α-Methyl-L-proline
4026653.0001
4026653.0005
F-3440.0001BAEA
571.91
USD
F-3440.0001BA
F-3440.0005BA
ɑ-Methyl-L-proline
alpha-Methyl-L-proline
Sequence: H-α-Me-Pro-OH
α-Methyl-proline restores normal levels of bone collagen type I synthesis in ovariectomized rats, used as models for human postmenopausal osteoporosis.
Formula:
C₆H₁₁NO₂ MW: 129.16 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD01318647 CAS Number: 42856-71-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...