L-Homocitrulline

Supplier: BACHEM AMERICAS INC

Synonyms: H-HoCit-OH, (S)-2-Amino-6-ureidohexanoic acid

4019080.0001 4019080.0005
F-2995.0001BAEA 583.98 USD
F-2995.0001BA F-2995.0005BA
L-Homocitrulline
L-Homocitrulline

Sequence: H-Homocit-OH


Hci derivatives.


Synonym(s): H-Lys(carbamoyl)-OH

Formula: C₇H₁₅N₃O₃
MW: 189.215 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00038143
CAS Number: 1190-49-4
EINECS: 214-722-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR