(S)-2-Amino-4-phenylbutanoic acid
Supplier: BACHEM AMERICAS INC
4005372.0005
4005372.0025
F-1610.0005BAEA
392.15
USD
F-1610.0005BA
F-1610.0025BA
(S)-2-Amino-4-phenylbutanoic acid
Sequence: H-Homophe-OH
Hph derivatives. Bachem also offers Fmoc-Hph-Wang resin, D-2005.
Synonym(s): (S)-2-Amino-4-phenyl-butyric acid
Formula:
C₁₀H₁₃NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD00002619 CAS Number: 943-73-7 EINECS: 213-403-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...