1-Methyl-L-histidine

Supplier: BACHEM AMERICAS INC

Synonyms: H-His(1-Me)-OH

4041045.0250 4041045.1000
E-3665.0250BAEA 245.07 USD
E-3665.0250BA E-3665.1000BA
1-Methyl-L-histidine
1-Methyl-L-histidine

Sequence: H-His(1-Me)-OH


The atoms of the imidazole ring of histidine can be numbered in two different ways. Bachem as well as organic chemists designate the position of N-τ as 1 and the position of N-π as 3, whereas biochemists generally number N-π, the nitrogen atom adjacent to the side chain, as 1, and N-τ as 3.


Synonym(s): H-His(τ-Me)-OH

Formula: C₇H₁₁N₃O₂
MW: 169.184 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00005295
CAS Number: 332-80-9
EINECS: 206-368-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR