Trt-Glu(OtBu)-OH · DCHA
Supplier: BACHEM AMERICAS INC
4033393.0001
4033393.0005
E-3650.0001BAEA
196.08
USD
E-3650.0001BA
E-3650.0005BA
Trt-Glu(OtBu)-OH · DCHA
Trt-Glu(OtBu)-OH · DCHA
Sequence: Trt-Glu(OtBu)-OH · DCHA
The derivatives of Glu-NH₂ are assembled in the "Isoglutamine" subfamily.
Formula:
C₂₈H₃₁NO₄·C₁₂H₂₃N MW: 626.88 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD03424223 CAS Number: 86967-51-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...