N-Methyl-L-threonine
Supplier: BACHEM AMERICAS INC
Synonyms:
N-Me-Thr-OH
4029013.0001
4029013.0005
E-3480.0001BAEA
571.91
USD
E-3480.0001BA
E-3480.0005BA
N-Methyl-L-threonine
N-Methyl-L-threonine
Sequence: N-Me-Thr-OH
Bachem also offers phosphothreonine derivatives.
Formula:
C₅H₁₁NO₃ MW: 133.147 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD02094308 CAS Number: 2812-28-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...