H-Tyr(tBu)-allyl ester·HCl

Supplier: BACHEM AMERICAS INC

Synonyms: H-Tyr(tBu)-allyl ester·HCl

4026774.0001 4026774.0005
E-3335.0001BAEA 196.08 USD
E-3335.0001BA E-3335.0005BA
H-Tyr(tBu)-allyl ester·HCl
H-Tyr(tBu)-allyl ester·HCl

Sequence: H-Tyr(tBu)-allyl ester · HCl


Besides a large selection of tyrosine derivatives, Bachem offers derivatives of O-alkylated tyrosines, thyronine, phosphotyrosine and sulfotyrosine. Bachem additionally offers H-[¹⁵N]Tyr-OH, E-3245.

Formula: C₁₆H₂₄ClNO₃
MW: 313.822 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00798675
CAS Number: 218938-62-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR