N-Acetyl-S-trityl-L-cysteine

Supplier: BACHEM AMERICAS INC

Synonyms: Ac-Cys(Trt)-OH

4018217.0001 4018217.0005
E-3030.0001BAEA 228.74 USD
E-3030.0001BA E-3030.0005BA
N-Acetyl-S-trityl-L-cysteine
N-Acetyl-S-trityl-L-cysteine

Sequence: Ac-Cys(Trt)-OH

Formula: C₂₄H₂₃NO₃S
Storage Temperature: Refrigerator
MDL Number: MFCD00236747
CAS Number: 27486-87-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR