N'-Trityl-L-glutamine
Supplier: BACHEM AMERICAS INC
Synonyms:
H-Gln(Trt)-OH
4014947.0001
4014947.0005
4014947.0025
E-2815.0001BAEA
114.5
USD
E-2815.0001BA
E-2815.0005BA
E-2815.0025BA
N'-Trityl-L-glutamine
N'-Trityl-L-glutamine
Sequence: H-Gln(Trt)-OH
For isoglutamine derivatives please see the family "Glutamic Acid".
Formula:
C₂₄H₂₄N₂O₃ Storage Temperature: Ambient |
MDL Number:
MFCD00237060 CAS Number: 102747-84-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...