N-Methyl-L-proline

Supplier: BACHEM AMERICAS INC

Synonyms: N-Me-Pro-OH, H-N-Me-Pro-OH

4008358.0001 4008358.0005
E-2185.0001BAEA 212.39 USD
E-2185.0001BA E-2185.0005BA
N-Methyl-L-proline
N-Methyl-L-proline

Sequence: N-Me-Pro-OH

Formula: C₆H₁₁NO₂
MW: 129.159 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011565
CAS Number: 475-11-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR