N-Methyl-L-alanine

Supplier: BACHEM AMERICAS INC

Synonyms: H-N-Me-Ala-OH, N-Me-Ala-OH

4002601.0001 4002601.0005
E-2125.0001BAEA 114.5 USD
E-2125.0001BA E-2125.0005BA
N-Methyl-L-alanine
N-Methyl-L-alanine

Sequence: N-Me-Ala-OH


Bachem additionally offers deuterated alanine: Fmoc-[D₄]Ala-OH (B-4130).

Formula: C₄H₉NO₂
Storage Temperature: Ambient
MDL Number: MFCD00037241
CAS Number: 3913-67-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR