H-ORN(Z)-OH
Supplier: BACHEM AMERICAS INC
4001491.0025
4001491.0250
E-1720.0025BAEA
212.39
USD
E-1720.0025BA
E-1720.0250BA
H-ORN(Z)-OH
H-ORN(Z)-OH
Sequence: H-Orn(Z)-OH
Formula:
C₁₃H₁₈N₂O₄ Storage Temperature: Ambient |
MDL Number:
MFCD00037220 CAS Number: 3304-51-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...