O-tert-Butyl-L-tyrosine

Supplier: BACHEM AMERICAS INC

Synonyms: H-Tyr(tBu)-OH

4001967.0001 4001967.0005 4001967.0025
E-1685.0001BAEA 114.5 USD
E-1685.0001BA E-1685.0005BA E-1685.0025BA
O-tert-Butyl-L-tyrosine
O-tert-Butyl-L-tyrosine

Sequence: H-Tyr(tBu)-OH


Besides a large selection of tyrosine derivatives, Bachem offers derivatives of O-alkylated tyrosines, thyronine, phosphotyrosine and sulfotyrosine. Bachem additionally offers H-[¹⁵N]Tyr-OH, E-3245.

Formula: C₁₃H₁₉NO₃
MW: 237.30 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00037202
CAS Number: 18822-59-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR