Acetyl-L-threonine

Supplier: BACHEM AMERICAS INC

Synonyms: Ac-Thr-OH

4000580.0250 4000580.1000
E-1210.0250BAEA 114.5 USD
E-1210.0250BA E-1210.1000BA E-1210.5000BA
Acetyl-L-threonine
Acetyl-L-threonine

Sequence: Ac-Thr-OH


Bachem also offers phosphothreonine derivatives.

Formula: C₆H₁₁NO₄
MW: 161.157 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00037810
CAS Number: 17093-74-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR