PAL resin 200-400 mesh
Supplier: BACHEM AMERICAS INC
4031247.0001 4031247.0005
D-2125.0001BAEA 251.47 USD
D-2125.0001BA D-2125.0005BA
PAL resin 200-400 mesh
PAL (Peptide Amide Linker) resin
Useful for the synthesis of peptide amides using Fmoc strategy. Cleavage has been performed with 50 % TFA in CH₂Cl₂ or 95% aqueous TFA. Scavengers may be required.
  • Substitution: 0.2-0.5 mmol/g