(S)-4-Amino-2-(benzyloxycarbonylamino)butyric acid

Supplier: BACHEM AMERICAS INC

Synonyms: (S)-4-Amino-2-(((benzyloxy)carbonyl)amino)butanoic acid, Z-Dab-OH, Cbz-α-Dab-OH

4015635.0001 4015635.0005 4015635.0025
C-3705.0001BAEA 114.5 USD
C-3705.0001BA C-3705.0005BA C-3705.0025BA
(S)-4-Amino-2-(benzyloxycarbonylamino)butyric acid
(S)-4-Amino-2-(benzyloxycarbonylamino)butyric acid

Sequence: Z-Dab-OH

Formula: C₁₂H₁₆N₂O₄
Storage Temperature: Ambient
MDL Number: MFCD00077904
CAS Number: 62234-40-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR